Learn More
Schiff's Fuchsin-Sulfite Reagent, Sigma-Aldrich
Catalog No. 01258822 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-822 500 mL
01-258-887 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-258-822 Supplier MilliporeSigma Supelco Supplier No. S5133500ML
Add to Cart
Add to Cart
Schiff's fuchsin-sulfite reagent is a part of the glycoprotein detection kit. It is primarily used in the Periodic Acid-Schiff (PAS) staining method to detect and visualize aldehydes, glycoproteins in biological samples. The reagent consists of basic fuchsin dye and sodium bisulfite. It reacts with the aldehyde groups of carbohydrates and produces a pink to magenta color, allowing for the identification of these substances in samples.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 500 mL
Product Type Schiff's Reagent