Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LifeSecure™ SchoolGuard™ Easy-Roll Classroom Evacuation and Lockdown Kit
SDPSCI_ED

Catalog No. S29693 Shop All Life Secure Products
Change view
Click to view available options
Product Type:
BleedStop Compact 100 Bleeding Control Kit
BleedStop Compact 200 Bleeding Control Kit
Includes:
SOFT-W tourniquet
SWAT-T tourniquet
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Includes
S29693 BleedStop Compact 100 Bleeding Control Kit SWAT-T tourniquet
S29694 BleedStop Compact 200 Bleeding Control Kit SOFT-W tourniquet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S29693 Supplier LifeSecure™ Supplier No. 31855
Add to Cart
Add to Cart

This Easy-Roll high-visibility school emergency kit for up to 29 students and 1 teacher meets all 10 essential survival needs for lockdown or evacuation and includes a Bleeding Control Kit for treating bleeding wounds from accidents, disasters, or active shooter situations.

Includes:

  • MobileAid™ BleedStop Compact Bleeding Control Kit
  • Tourniquet
  • Mini-compress bandage
  • QuickClot blood clotting gauze
  • Petroleum gauze for chest wounds
  • Bleeding wound supplies

Portable fold-flat toilet stores in the survival backpack with other survival supplies: water, food, breathing protection, first aid, shelter, warmth, communication, light, tools, and sanitation/hygiene

Specifications

Product Type BleedStop Compact 100 Bleeding Control Kit
For Use With (Application) Lockdown and evacuation
Length (Metric) 35.56 cm
Color Blue, Orange
Includes SWAT-T tourniquet
Quantity 1 Ea.
Sufficient For 29 Students and 1 Teacher
Dimensions (L x W x H) 14 x 13 x 21 in.
Width (Metric) 33.02 cm
Height (Metric) 53.34 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.