Learn More
Strem, An Ascensus Company CAS 329735-77-5. 25mg. 26-Diisopropylphenylimidoneophylidene[(R)-()-BIPHEN]molybdenum(VI) min. 97% (R) SCHROCK-HOVEYDA CATALYST. MFCD01862462
SDP
Supplier:  Strem, An Ascensus Company 42121325MG
Encompass_Preferred
CAS 329735-77-5. 25mg. 26-Diisopropylphenylimidoneophylidene[(R)-()-BIPHEN]molybdenum(VI) min. 97% (R) SCHROCK-HOVEYDA CATALYST. MFCD01862462. Molecular Weight 755.93. Molecular Formula Mo(C10H12)(C12H17N)(C24H32O2). Color/form red xtl. Strem 42-1213. http//www.strem.com/catalog/v/42-1213/
Catalog No. 50-211-0815
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.