Learn More
Thomas Scientific 96 Pin Sterile Replicator, Standard Length, 120/Cs, 12mm Long, 1mm Diameter, Packaged 10/Bag
Supplier: Thomas Scientific 1149C50
The 96 Pin Replicator is a disposable pin replicator for one time use, that virtually eliminates the threat of cross-contamination from repeat use metal replicators.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.