Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Science Fair Kits - Film Canister Rockets
SCI_ED

Catalog No. S30768 Shop All Eisco Products
Change view
Click to view available options
Class Size:
25 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S30768 25 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S30768 Supplier Eisco™ Supplier No. GP00028
Add to Cart
Add to Cart

Launch a rocket using vinegar and Alka-Seltzer. The Film Canister Rocket Kit includes enough to build and launch your own rocket, and explore mass, measurement, and simple chemical reactions.

  • Combine the ingredients into the film canister and watch as the canister launches into the air
  • Team up with your friends to see whose rocket can launch the highest
  • Simple project for fun, hands on learning
  • Kit includes:
    • (2) Film Canisters
    • (1) 10 mL Bottle of Vinegar
    • (2) Packs of Alka-Seltzer tablets (2 tablets per pack)
    • Experiment Instructions
    • Chemical Safety Sheet
  • Safety goggles are required, not included

Specifications

Grade 3 to 12
Age 8 to 18
For Use With (Application) Engineering
Includes (2) Film Canisters, (1) 10 mL Bottle of Vinegar, (2) Packs of Alka-Seltzer tablets (2 tablets per pack), Experiment Instructions, Chemical Safety Sheet
Material Plastic, Paper, Polymer
Quantity 1 Ea.
Class Size 25 Students
Product Type Activity Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.