Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Science Fair Kits - Light Sensitive Bristlebot Kit
SCI_ED

Catalog No. S30767 Shop All Eisco Products
Change view
Click to view available options
Age:
10 to 18
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Age
S30767 10 to 18
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S30767 Supplier Eisco™ Supplier No. GP00022

missing translation for 'validMainframeMsg'

Explore a simple feedback circuit analog of bacteria. The Light Sensitive Bristlebot Kit includes components for building a simple light-based bacteria model based on the bristlebot design.

  • Explore a simple feedback circuit analog of bacteria.
  • The bot moves randomly when there isn't enough light. When it wanders into a region with enough light it slows down and stops
  • Scissors required, not included
  • Kit includes:
    • (2) toothbrush heads
    • (1) 25 pin micro breadboard
    • (1) 2N3904 NPN bipolar transitor,
    • (1) CR2032 3V Lithium coin cell battery
    • (1) Micro 3V DC vibration motor
    • (1) 5539 photoresistor
    • mounting tape
    • single-use superglue
    • (4) googly eyes
    • (2) bare metal paperclips
    • detailed, illustrative instructions, and pedagogical materials
  • Team up multiple bots to explore group behavior
  • Exploration of the kit takes about an hour, including assembly

Specifications

Grade 5
Age 10 to 18
For Use With (Application) Engineering
Includes (2) toothbrush heads, (1) 25 pin micro breadboard, (1) 2N3904 NPN bipolar transitor, (1) CR2032 3V Lithium coin cell battery, (1) Micro 3V DC vibration motor, (1) 5539 photoresistor, mounting tape, single-use superglue, (4) googly eyes, (2) bare metal paperclips, detailed, illustrative instructions, and pedagogical materials
Material Metal, plastic, polymer, rubber
Quantity 1
Product Type Activity Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.