Description
- Two cubes to our 10-cube set
- Lignum Vitae (ironwood), one o fthe heavies and hardest woods in the world, sinks in water, although because it is wood, you students may expect it to float
- Polypropylene floats in water, although because it is dense plastic, your students may expect it to sink
- 12 one-inch cubes, with instructions and handy storage case
Specifications
Specifications
| Product Type | Cube Set |
| For Use With (Application) | Demonstrate density experiment in a dozen ways |
| Includes | Instructions and handy storage case |