Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Science First™ Heros Fountain
SDPSCI_ED

Catalog No. S98899 Shop All Science First Products
Change view
Click to view available options
Includes:
Instructions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
S98899 Instructions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S98899 Supplier Science First™ Supplier No. 6112250
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Demonstrates fluid pressure.

  • Spouts water into the air for several minutes
  • As water in the top enters the lower bulb, it forces the air in the bulb upward, compressing more air in the upper bulb
  • The water has nowhere to go but out the upper tube
  • With instructions

Order Info

Height: 50cm

Specifications

Includes Instructions
Product Type Hero's Fountain
Height (Metric) 50 cm
For Use With (Application) Demonstration of fluid pressure spouts water into the air for several minutes
Height (English) 19.6 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.