Learn More
Science First™ Magnetic Earth Model
SDPSCI_ED
Catalog No. S99629 Shop All Science First Products
Change view
Click to view available options
Magnetic earth model
Catalog No.
S99629 Magnetic earth model
1 options
Catalog No. S99629 Supplier Science First™ Supplier No. 6539015
This is a great demonstration of the earth's magnetic field.
  • Used as it is or place it on an overhead projector to show the whole class
  • Iron filings are suspended in oil to allow the included magnet to align them to its field
Specifications
For Use With (Application) To demonstrate Earth's magnetic field
Product Type Magnetic Earth Model