Description
Includes
One mole (6.03 x 1023 particles) of aluminum, copper, iron, zinc, plus instructions
- Similar to Mole Box, an 11 in. cube which shows 22.4l (one mole) of air
- Our specimen set shows the same concept in solid form
Specifications
Specifications
| For Use With (Application) | To show one mole (22.4l) of air in solid form, 11 in. cube |
| Includes | One mole (6.03 x 1023 particles) of aluminum, copper, iron, zinc and instructions |
| Product Type | Mole Specimen Set |