Learn More
Description
Includes
Instruction manual
Model demonstrates how a jar filled with water shoots out at different rates of speed. The water at the bottom will have the most water above it, increasing the pressure and hence the rate of speed at which it spouts. The water pressure decreases proportionately for the two upper nipples. If the pressure is low, water won't escape at all.
- Unbreakable polycarbonate tube
- Color: clear
- Three brass nipples positioned one on top of each other
- Mounted on stand
Specifications
Specifications
| Product Type | Perforated Jar |
| Includes | Instruction manual |
| For Use With (Application) | Show how a fluid changes behavior even under it's own weight |
| Subject | Physics |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

