Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NewPath Learning™ Science Visual Learning Guides
SDPSCI_ED

Catalog No. S01885 Shop All United Scientific Supplies, Inc Products
Catalog No. S01885 Supplier NewPath Learning™ Supplier No. 33600
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Use visual learning methods to improve student performance.

UniStore™ Wide Mouth Square Bottles are available in different materials and sizes for multiple applications. The high-density polyethylene bottles (HDPE) have excellent chemical resistance to most acids, bases, and alcohols. The square design allows the bottles to nest together for more efficient storage, packing and shipping. These HDPE bottles are leakproof and have a polypropylene cap.

  • Square design for compact storage
  • Excellent chemical resistance
  • Precision molded for consistent wall thickness with smooth interior and exterior surfaces
  • FDA-21CFR compliant resin
  • Leakproof closure
  • Available in 60 mL, 175 mL, 250 mL, 500 mL, and 1000 mL

Specifications

Capacity (Metric) 60 mL
Color Translucent White
Material HDPE
Sterility Non-sterile
Closure Included Yes
Graduated No
Mouth Wide, Threaded
Autoclavable Not Autoclavable
Capacity (English) 2 oz.
Product Type Reagent Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.