Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NewPath Learning™ Science Visual Learning Guides
SDPSCI_ED

Catalog No. p-3973425 Shop All United Scientific Supplies, Inc Products
Catalog No. S01887 Supplier NewPath Learning™ Supplier No. 33605
Only null left
Add to Cart
Add to Cart

Use visual learning methods to improve student performance.

UniStore™ Wide Mouth Square Bottles are available in different materials and sizes for multiple applications. The square design allows the bottles to nest together for more efficient storage, packing and shipping. The polypropylene copolymer (PPCO) bottles are autoclavable at 121°C. These bottles are leakproof and have a polypropylene cap.

  • Square design for compact storage
  • Better impact strength and durability than standard polypropylene (PP)
  • Precision molded for consistent wall thickness with smooth interior and exterior surfaces
  • FDA-21CFR compliant resin
  • Autoclavable at 121°C
  • Leakproof closure
  • Available in 60 mL, 175 mL, 250 mL, 500 mL, and 1000 mL

Specifications

Capacity (Metric) 60 mL
Capacity (English) 2 oz.
Color Translucent White
Material Plastic
Sterility Non-sterile
Closure Included Yes
Graduated No
Mouth Wide, Threaded
Product Type Reagent Bottle
Autoclavable Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.