Learn More
Research Products International Corp Magnetic Stirrer, 8 x 8 Inch Surface
Supplier: Research Products International Corp 437042
EchoTherm Single Stirring Position Analog Stirrer
Features:
- Stir from 100 to 1500 rpm on each stirring position with individual controls
- Stir 2 liters water
- Fused for safety
- Rugged cast aluminum chassis
- 12-month warranty
- UL, CSA and CE certified
- Magnet strength 20 lbs