Learn More
Scientific Device Laboratory Disposable Forceps
SDP
Catalog No. 23769542 Shop All Scientific Device Laboratory Inc. Products
Change view
Click to view available options
Length (English):
4.5 in.
Catalog No. Length (English)
23-769-542 4.5 in.
1 options
This item is not returnable. View return policy
Catalog No. 23-769-542 Supplier Scientific Device Supplier No. 900
This item is not returnable. View return policy
With sturdy gripping surface
Specifications
Product Type Sterile Forceps
Length (English) 4.5 in.
Material Stainless Steel
No. per Pack 25
Tip Style Grooved