Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scientific Device Snap n' Digest™ Digestion System
SDP

Catalog No. 23769318 Shop All Scientific Device Laboratory Inc. Products
Change view
Click to view available options
Quantity:
10 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-769-318 10 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-769-318 Supplier Scientific Device Supplier No. 668

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Ten non-breakable bottles, five packets of buffer

Simple and inexpensive fluid processing of mycobacterial specimens

  • Bottles contain an alkali solution and a sealed glass ampoule of pre-weighed NALC
  • Glass ampoule breaks easily when needed and the contents rapidly dissolve in fluid
  • Bottles are equipped with a safety tube to help trap and break NALC capsule
  • Self-contained system
  • Fill size: 150mL (5.1 oz.)

Specifications

Product Type Snap N' Digest
Capacity (English) 5.1 oz.
Capacity (Metric) 150 mL
For Use With (Application) For processing mycobacterial specimens
Quantity 10 Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.