Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Scientific Glucose Test Strips - Educational Glucose Test Strips - 100 Strips
SCI_ED

Catalog No. S02915 Shop All Eisco Products
Change view
Click to view available options
Quantity:
100 Strips
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S02915 100 Strips
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S02915 Supplier Eisco™ Supplier No. GLUTEST100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Eisco Labs scientific glucose test strips are essential tools for food science and osmosis/diffusion experiments. Ideal for classroom and lab use, providing accurate and reliable results for various scientific investigations and demonstrations.

  • Glucose meter strips detect glucose levels from 0 to 3000 mg/dL or 0 to 3% ensuring precise results vital for educational experiments in various disciplines
  • Test Strips are ideal for measuring glucose levels in osmosis/diffusion experiments and food science studies, offering insights into glucose behavior in various solutions
  • The color chart features blocks at 0, 100, 300, 1000, and 3000 mg/dL, ensuring accurate and easy-to-read results for educational and scientific purposes
  • Utilizes an enzyme specific for glucose detection. Note: Mutarotation of glucose in water samples may require up to 3 hours for optimal enzyme function
  • The glucose test kit is intended for scientific and educational purposes only.

Specifications

Clia Complexity Not Applicable
Concentration 0 to 3000 mg/dL
Detectable Analytes Glucose
Format Test Strip
Type Glucose Detection
For Use With (Application) Glucose Detection
Quantity 100 Strips
Form Ready-to-Use (RTU)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.