Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Globe Scientific Scintillation Vials

Catalog No. 220101157
Change view
Click to view available options
Volume (Metric):
4 mL
6.5 mL
20 mL
Height (Metric):
53 mm
57 mm
61.4 mm
Diameter (Metric) Outer:
13.8 mm
16 mm
26.5 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Height (Metric) Diameter (Metric) Outer
22-010-1157 4 mL 53 mm 13.8 mm
22-010-1156 20 mL 61.4 mm 26.5 mm
22-010-1158 6.5 mL 57 mm 16 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-010-1157 Supplier Globe Scientific Supplier No. 101030
Add to Cart
Add to Cart
This item is not returnable. View return policy

Designed to provide consistently low background counts and are suitable for low activity counting.

20mL Scintillation Vial:

  • Scintillation counting, gamma counting, chromatography, sample storage and culturing
  • Because of its leak-tight cap it is also commonly used to store and transport histological specimens in 10% formalin
  • Provides high degree of resistance to organic solvents and sufficient translucence so that reagent levels can be easily seen
  • Unique thread design of closure permits vial to be sealed tight with easy 1/4 turn and with minimum pressure

6.5mL Scintillation Vial:

  • Securely sealed by simply snapping caps on
  • Cap is conveniently removed with easy 1/4 turn to left

4mL Polyethylene Scintillation Vial:

  • Uses full turn polyethylene screw cap
  • Most economical size to use because it requires least amount of solvent

Specifications

Diameter (Metric) Outer 13.8 mm
Material HDPE
Sterility Non-sterile
Volume (Metric) 4 mL
No. per Case 2000
Closure Material Polyethylene
Closure Type Screw Cap
Height (Metric) 53 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.