Learn More
Roboz Surgical SCISSORS-BLUNT CURVED 6 1/2
SDP
Supplier:  Roboz Surgical RS6943
Encompass
Veterinary Scissors; Heavy Pattern; Curved, Blunt-Blunt Serrated Blades; Blade Length 57mm 6-1/2 in. Overall Length. Veterinary Scissors have one smooth blade and one serrated blade which allows for a sharp cut through softer bone and cartilage.
Catalog No. NC0403760
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.