Learn More
World Precision Instrument Noyes Scissors, 14 cm, Straight, Sharp/Sharp, 20 mm Blades
SDP
Supplier:  World Precision Instrument 501237
Encompass
Noyes Micro Scissors are a surgical instrument designed for ophthalmic surgery and are an ideal choice for microdissection or for delicate small animal surgeries. Named after their inventor, ophthalmologist Dr. Arthur Noyes, the Noyes Micro Scissors are characterized by their small size and fine tips. They let you make precise and controlled incisions. These surgical scissors are typically used for delicate tasks such as cutting fine structures, trimming tissue, or creating small openings. Noyes scissors are a type of spring scissors which can be used ambidextrously. For this reason, Noyes scissors are often preferred by left-handed surgeons. These scissors are 14 cm (5.5") long and come with straight or curved tips. Noyes Micro Scissors are made from surgical grade stainless steel, which is suitable for autoclaving or chemical sterilization. Always clean your surgical instruments promptly after you use them.
Catalog No. 50-822-583