Learn More
Med Vet International Lister Bandage Scissors, 3.5 in., Economy
Supplier:  Med Vet International 2120
Encompass_Preferred
Lister bandage scissors are commonly used for sizing dressings and removing circumferential bandages. They may also be used to cut tougher material such as plaster, fabric, and occasionally umbilical cord. The jaws of the are angled with the lower blade being slightly longer.
Catalog No. 50-222-4024
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.