Learn More
AdipoGen Sclerotiorin (1 mg)
SDP
Supplier:  AdipoGen AGCN20054M001
Encompass_Preferred
Chemical. CAS: 549-23-5. Formula: C21H23ClO5. MW: 390.9. Isolated from fungus Penicillium sp. Antibiotic. Antibacterial. Weak antagonist of human endothelin receptor A (ETA) and B (ETB). Cholesteryl ester transfer protein (CETP) activity inhibitor. Grb2-Shc binding inhibitor. Potent aldose reductase inhibitor. Potent reversible, non-competitive soybean lipoxygenase-1 (LOX-1) inhibitor. Antioxidant. Free radical scavenger. Antifungal.
Catalog No. 50-205-3397
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.