Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCMH1 Monoclonal Antibody (OTI5E6), TrueMAB™, OriGene
SDP

Catalog No. 501682148
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-2148 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-2148 Supplier OriGene Supplier No. TA808456
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Component of the Polycomb group (PcG) multiprotein PRC1 complex, a complex required to maintain the transcriptionally repressive state of many genes, including Hox genes, throughout development. PcG PRC1 complex acts via chromatin remodeling and modification of histones; it mediates monoubiquitination of histone H2A ′Lys-119′, rendering chromatin heritably changed in its expressibility.

Specifications

Antigen SCMH1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI5E6
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene SCMH1
Gene Accession No. Q96GD3
Gene Alias Scml3
Gene Symbols SCMH1
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 1-309 of human SCMH1 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22955
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.