Learn More
Invitrogen™ SCO2 Polyclonal Antibody

Description
Immunogen sequence: PTAWHRLSQL KPRVLPGTLG GQALHLRSWL LSRQGPAETG GQGQPQGPGL RTRLLITGLF GAGLGGAWLA LRAEKERLQQ QKRTEALRQA AVGQGDFHLL DHRG Highest antigen sequence identity to the following orthologs: Mouse - 65%, Rat - 26%.
Specifications
Specifications
| Antigen | SCO2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Sco2 |
| Gene Accession No. | O43819 |
| Gene Alias | CEMCOX1; MYP6; protein SCO2 homolog, mitochondrial; SCO cytochrome oxidase deficient homolog 2; SCO cytochrome oxidase deficient homolog 2 (yeast); SCO1L; SCO2; SCO2 cytochrome c oxidase assembly protein |
| Gene Symbols | Sco2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.