Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex Scope Cabinet - Wall Mount
SDP

Catalog No. 01259219
Change view
Click to view available options
Cabinet Type:
Double Scope
Large
Single Scope
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cabinet Type
01-259-219 Large
01-259-131 Double Scope
01-259-218 Single Scope
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
N/A
Catalog No. 01-259-219 Supplier Poltex Supplier No. SCOPHNG2LGW
Get a Poltex Pipette Holder with $3,000 Purchase
View all promotions
Add to Cart
Add to Cart
N/A

Endoscope storage cabinet with door mounted on stainless steel hinges and a magnetic catch for secure closing. Wall mount design with durable and cleanable PETG construction.

  • Endoscope storage cabinet.
  • The door is mounted with stainless steel hinges and has a magnetic catch for secure closing.
  • Choose from several different options.
  • Wall Mount (screws/anchors included).
  • Durable and cleanable PETG construction.
  • Consumables not included.

Specifications

Product Type Storage Cabinet
Warranty 90 Day
Weight (English) 8 lb.
For Use With (Application) Endoscope Storage Cabinet
Door Type PETG, Magnetic Catch
No. of Shelves 1
No. of Doors 1
Hinge Side Left
Color Clear
Dimensions (L x W x H) 4.9 x 8.3 x 40 in.
Cabinet Type Large
Material PETG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.