Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scopolamine Methyl Nitrate 98.0+%, TCI America™
SDP

Catalog No. S02301G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S02301G 1 g
S02305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S02301G Supplier TCI America Supplier No. S02301G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6106-46-3
Molecular Formula C18H24N2O7
Molecular Weight (g/mol) 380.40
MDL Number MFCD00078239
InChI Key BSQIVYOSLFLSGE-ODDNLWGVSA-N
Synonym Methscopolamine Nitrate
PubChem CID 24847384
IUPAC Name (1S,2R,4S,5S)-7-[(3-hydroxy-2-phenylpropanoyl)oxy]-9,9-dimethyl-3-oxa-9-azatricyclo[3.3.1.0²,⁴]nonan-9-ium nitrate
SMILES [O-][N+]([O-])=O.C[N+]1(C)[C@H]2CC(C[C@H]1[C@@H]1O[C@H]21)OC(=O)C(CO)C1=CC=CC=C1

Specifications

Melting Point 199°C
Color White
Quantity 1 g
UN Number 1544
Formula Weight 380.40
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Scopolamine Methyl Nitrate
EINECSNumber (5)-3899
RTECSNumber YM3675150
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.