Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Scotch™ Fine Line Tape

Catalog No. 19072017 Shop All 3M Products
Change view
Click to view available options
Quantity:
144 Rolls/Cs. (12 Rolls/Pk. 12 Pk./Cs.)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-072-017 144 Rolls/Cs. (12 Rolls/Pk. 12 Pk./Cs.)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-072-017 Supplier 3M™ Supplier No. 7000048455
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3M™ Scotch™ Fine Line Tape is a high-performance polypropylene plastic tape that has both a low profile and high adhesion for use in painting long straight lines and sweeping curves where a crisp, sharp paint line is desired.

  • Thin tape provides excellent color line separation
  • Proven excellent for painting long straight lines and smooth curves
  • Resistant to most solvents and moisture
  • Often used as a layout tape for high value multi-color and custom painted graphics.
TRUSTED_SUSTAINABILITY

Specifications

Color Green
Material Rubber
Product Type Fine Line Tape
For Use With (Application) Masking for Painting, Paint Preparation Masking
Model 218
Quantity 144 Rolls/Cs. (12 Rolls/Pk. 12 Pk./Cs.)
Thickness (English) 5 mil
Dimensions (L x W) 60 yd. x 1/8 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.