Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Scotch™ Fine Line Tape

Catalog No. 19070921 Shop All 3M Products
Change view
Click to view available options
Quantity:
24/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-070-921 24/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-070-921 Supplier 3M™ Supplier No. 7000028860
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3M Company Scotch™ Fine Line Tape has both a low profile and high adhesion for use in painting long straight lines and sweeping curves where a crisp, sharp paint line is desired

  • Resistant to most solvents and moisture
  • Thin tape provides excellent color line separation
  • 3M Catalog# 218

TRUSTED_SUSTAINABILITY

Specifications

Color Green
Material Polypropylene
Product Type Masking and Duct Tapes
For Use With (Application) Painting and Depainting
Model 218
Quantity 24/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.