Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Scotch™ Flatback Masking Tape

Catalog No. 19072046 Shop All 3M Products
Change view
Click to view available options
Quantity:
36 Rolls/Cs. (6 Rolls/Pk., 6 Pk./Cs.)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-072-046 36 Rolls/Cs. (6 Rolls/Pk., 6 Pk./Cs.)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-072-046 Supplier 3M™ Supplier No. 7000001488
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Scotch™ Flatback Masking Tape is used as a quality control paint adhesion testing tape.

  • Clean removal from paint, plating and powder coatings
  • Aggressive, pressure sensitive adhesive provides instant adhesion
  • Strong flatback paper backing resists tearing and breakage
  • Controlled unwind and easy to position, manipulate and tear by hand
  • High peel adhesion
  • Ideal for bundling and wrapping applications.
TRUSTED_SUSTAINABILITY

Specifications

Color Tan
Material Rubber
Product Type Masking Tape
For Use With (Application) Automotive, Design and Construction, Manufacturing, Mining, Oil and Gas, Transportation
Model 250
Quantity 36 Rolls/Cs. (6 Rolls/Pk., 6 Pk./Cs.)
Thickness (English) 6 mil
Dimensions (L x W) 60 yd. x 1 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.