Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Scotch™ High Performance Packing Tape

Catalog No. 19072090 Shop All 3M Products
Change view
Click to view available options
Dimensions (L x W):
50 m x 48 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19-072-090 50 m x 48 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-072-090 Supplier 3M™ Supplier No. 7010354113

Scotch™ Box Sealing Tape 313 is a high performance packing tape that is suitable for hard to stick surfaces and cold temperature applications

  • High performance packaging tape suitable for hard to stick surfaces and cold, damp environments
  • High coating weight acrylic adhesive with polypropylene backing delivers dependable, quality performance
  • Ideal for specialty, difficult and extreme applications
  • Resists UV rays without yellowing for archival applications
  • Meets FDA guidelines for indirect food contact
  • High edge tear and split resistance
  • Excellent adhesion to recycled liner paper and fiberboard
  • Resists abrasion, moisture, chemicals and scuffing for long-lasting performance
  • Polypropylene film backing is easy to handle and resists abrasion, moisture and scuffing.
  • Consistent and reliable backing easily conforms around edges and on rough surfaces for a tight seal.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Box Sealing Tape
For Use With (Application) Automated Carton Sealing, Box Closure, Cold Storage and Tamper Evident
Adhesive Type Acrylic
Color Clear
Description High performance
Dimensions (L x W) 50 m x 48 mm
Material Acrylic
Model 313
Quantity 36/Cs.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.