Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Scotch™ Masking Tape for Hard-to-Stick Surfaces

Catalog No. 19072108 Shop All 3M Products
Change view
Click to view available options
Quantity:
24/Cs.
36/Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-072-108 24/Cs.
19-072-107 36/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-072-108 Supplier 3M™ Supplier No. 7000126658
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3M™ Scotch™ Masking Tape for Hard-to-Stick Surfaces can be removed cleanly for up to 3 days

  • For use on brick, concrete, rough wood and stucco
  • Can be used with lacquer coatings
  • Very High Adhesion level
  • Bonds aggressively, yet has a smooth unwind
TRUSTED_SUSTAINABILITY

Specifications

Color Green
Material Rubber
Product Type Masking Tape
For Use With (Application) Consumer, Design and Construction
Model 2060-48A-BK
Quantity 24/Cs.
Dimensions (L x W) 55 m x 48 mm (60.1 yd. x 1.88 in.)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.