Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Scotch™ Ultimate Paint Edge Masking Tape

Catalog No. 19072084 Shop All 3M Products
Change view
Click to view available options
Quantity:
24/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-072-084 24/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-072-084 Supplier 3M™ Supplier No. 7100106569
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Scotch™ Ultimate Paint Edge Masking Tape features unique acrylic adhesive technology for good, instant adhesion

  • Temperature-resistant, capable of withstanding paint bake operations of up to 300°F
  • Peels off cleanly, without leaving adhesive residue
  • Ultra-thin backing provides a low paint ridge for sharp, smooth paint lines
  • Adhesive maintains good holding power that resists lifting and curling
  • Acrylic adhesive provides UV resistance, which allows outdoor use for up to 21 days with clean removal from most surfaces
  • Solvent and moisture resistance prevents paint bleedthrough
TRUSTED_SUSTAINABILITY

Specifications

Color Gold
Material Acrylic
Product Type Edge Masking Tape
For Use With (Application) Automotive, Design and Construction, Manufacturing, Transportation
Model 2460
Quantity 24/Cs.
Thickness (English) 3.3 mil
Dimensions (L x W) 60 yd. x 2 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.