Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ ScotchBlue™ Painter's Tape Display

Catalog No. 19072114 Shop All 3M Products
Change view
Click to view available options
Quantity:
54 Rolls/Display
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-072-114 54 Rolls/Display
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-072-114 Supplier 3M™ Supplier No. 7010337159
This item is not returnable. View return policy

3M™ ScotchBlue™ Painter's Tape Display is a medium adhesion tape that is ideal for painted walls and trim, woodwork, glass and metal.

  • Ideal for use on smooth or lightly textured walls, trim, glass and metal
  • Sharp paint lines on a variety of surfaces
  • Removes cleanly up to 14 days with no sticky residue, even in direct sunlight
  • Medium adhesion level removes easily, while protecting your surfaces
  • Indoor or outdoor use
  • For ideal paint results make sure the surface is clean, dry and dust-free so that the tape sticks properly. Next apply tape onto the surface while pressing down as you go, then secure the tape by pressing down on it firmly with a smooth edge. And lastly let the tape set for about 30 to 60 minutes before painting. Wait until the paint is dry to the touch before removing the tape. Lift it up by slowly pulling it back on itself, then removing at a 45 degree angle.
TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Product Type Painter's Tape
For Use With (Application) Decorating, Organizing and Crafts, DIY, Home Improvement, Home, Office, School Supplies
Model 2090-48B-N
Quantity 54 Rolls/Display
Dimensions (L x W) 60 yd. x 1.88 in. (54.8 m x 48 mm)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.