Learn More
Scott Safety™ Air-Pak™ X3 SCBA
Catalog No. 15499132
Change view
Click to view available options
4.5 (4500 psi); Gauge and HUD only; Standard hose
Catalog No.
15-499-132 4.5 (4500 psi); Gauge and HUD only; Standard hose
1 options
This item is not returnable. View return policy
Catalog No. 15-499-132 Supplier Scott Safety™ Supplier No. X3314021200101
This item is not returnable. View return policy
Meets requirements outlined in the 2013 Edition of the 1981 and 1982 standards.
Designed with enhancements to comfort, hose and wire management, weight distribution, fire ground communications and serviceability.
  • Fire ground communications enhanced with the introduction of the AV-3000 HT facepiece, EPIC 3 Communication and optimally positioned “buddy” lights
  • “External” HUD for ease in air status updates of the team
  • Dual redundant pressure reducer
  • Vibralert tactile alarm
Accountability features available on selected models:
  • Pak Alert PASS (Personal Alert Safety System) - standardized sound signals man-down situation
  • Pak Tracker Firefighter Locator Systems with HHR (handheld receiver) - alerts remote users of man-down situation; directional HHR provides direction for rescue teams
  • SEMS II - includes Pak Alert PASS and PAK Alert locator features plus redundant network to carry PASS and low-air notifications, reducing radio traffic
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance 1981, 2007, 2013 NFPA
Product Type SCBA
Pressure 4500 psi
For Use With (Application) Firefighting
Harness Style Standard
Hose Type Standard hose
Includes Gauge and HUD
Model X3314021200101