Scott Safety™ AV-2000™ Facepiece

The original Scott facepiece, the AV-2000 facepiece was first designed for maximum versatility

Manufacturer: Scott Health & Safety 80525728

 View more versions of this product

Catalog No. 19-827-919

  Check Availability
Add to Cart

Required Accessories : To use as an air-purifying respirator, requires cartridge adapter 805622-01 for 742 Series cartridges, 805059-01 for 40mm NIOSH cartridges and 200423-01 for CBRN Cap-1 canister.

    Specifications

    For Use With (Equipment) Av-2000™ facepiece
    Product Type Facepiece
    For Use With (Application) To minimize equipment inventory, simplify user training, reduce redundant fit testing, improved communications in the workplace
    Includes Speaking diaphragm, dual voicemitter, grey nosecup
    Size One
    Certifications NIOSH, ANSI Z87.1-2003, NFPA 1981:2007
    Harness Style Rubber
    Color Black

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit