Learn More
Scott Safety™ AV-2000™ Facepiece
Description
Requires
To use as an air-purifying respirator, requires cartridge adapter 805622-01 for 742 Series cartridges, 805059-01 for 40mm NIOSH cartridges and 200423-01 for CBRN Cap-1 canister.
- Top-Down Convertibility allows a single facepiece for APR, PAPR, SAR and SCBA respirators to reduce cost of ownership
- One fit test and reduced training
- Comfort Seal™ facepiece features a deep chin cup for reliable, comfortable fit
- Mechanical and electronic communication options across all applications.
- Kevlar™, polyester or rubber head harness options available in four color-coded sizes
Order Info
Fit testing required prior to using this facepiece. Prescription lens kit (804442-01) and other accessories available separately.
Certifications
All head harnesses NIOSH approved for APR, supplied air, SCBA and PAPR respirators. Rubber and Kevlar head harnesses also NIOSH approved for CBRN respirators.
Compliance
All head harnesses meet ANSI Z87.1-2003. Rubber and Kevlar head harnesses also meet NFPA 1981:2007.
Specifications
Specifications
| Certifications/Compliance | NIOSH, ANSI Z87.1-2003 |
| Color | Black |
| Material | Polyester, EPDM Rubber Faceshield |
| Product Type | Facepiece |
| Format | Large |
| For Use With (Application) | To minimize equipment inventory, simplify user training, reduce redundant fit testing, improved communications in the workplace |
| For Use With (Equipment) | Av-2000™ facepiece |
| Includes | Speaking diaphragm, dual voicemitter, gray nosecup |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.