Learn More
Scott Safety™ AV-3000™ Facepiece with SureSeal
Description
Requires
To use as an air-purifying respirator, requires cartridge adapter 805622-01 for 742 Series cartridges, 805059-01 for 40mm NIOSH cartridges and 200423-01 for CBRN Cap-1 canister.
- One fit test and reduced training
- Patent-pending SureSeal technology for the most advanced fit and comfort
- Wide seal offers a more secure interface with protective suits, hoods, shrouds and helmets
- Mechanical and extensive electronic communication options across all applications
- All components are latex free
- Kevlar™, polyester or rubber head harness options with three nosecup sizes
All head harnesses meet ANSI Z87.1-2003. Rubber and Kevlar head harnesses also meet NFPA 1981:2007. All head harnesses NIOSH approved for APR, supplied air, SCBA and PAPR respirators. Rubber and Kevlar head harnesses also NIOSH approved for CBRN respirators.
Order Info
Fit testing required prior to using this facepiece. Prescription lens kit (805753-01) and other accessories available separately.
Specifications
Specifications
| Certifications/Compliance | NIOSH, ANSI Z87.1-2003, NFPA 1981:2007 |
| Material | Dupont™ Kevlar™ harness, Polyisoprene Seal |
| Product Type | Facepiece |
| Format | Large |
| For Use With (Equipment) | Av-3000™ facepiece |
| Includes | Large dual voicemitters, communication bracket |
| Latex Free | Yes |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.