Learn More
Scott Safety™ Manifold Kits for Life-Link Emergency Breathing Support Systems
Catalog No. SCT2754002
Change view
Click to view available options
For Use With (Equipment):
Air-pak 2.2 and Air-pak Fifty SCBAS
Air-pak 4.5 and Air-pak Fifty SCBAS
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
SCT2754002 Air-pak 2.2 and Air-pak Fifty SCBAS
SCT2754004 Air-pak 4.5 and Air-pak Fifty SCBAS
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SCT2754002 Supplier Scott Safety™ Supplier No. 7012472571
Add to Cart
Add to Cart
Manifold kits provide fast, easy coupling during rescue operation so that two users of similarly equipped Air-pak™ system can share a common air supply.
For Air-Pak 2.2 and Air-Pak 4.5
Order Info
The items below must be installed by a Fisher Scientific™ Representative familiar with the procedures required to add an EBSS to an existing Air-Pak SCBA. For installation charges, call 1-800-772-6733.
Specifications
For Use With (Equipment) Air-pak 2.2 and Air-pak Fifty SCBAS
Model 2754002
Product Type Manifold Assembly