Learn More
Scott Safety™ SKA-PAK™ Supplied Air Respirators
Offers emergency respiratory protection for variety of applications.
Supplier: Scott Safety™ 7012623426
Description
- Designed for use in hazardous atmospheres
- Combination Type C positive pressure respirator with escape cylinder for entry into/escape from hazardous, confined space or IDLH situations
- Includes E-Z Flo pressure demand regulator with first-breath activation and low exhalation
- 5-, 10-, or 15-minute rated cylinder egress options for selection versatility based on task at hand
- Hip-mounted cylinder with durable, secure cylinder band retention system
- Available in two harness styles:
- Polypropylene - chemically resistant and lightweight
- Kevlar - Forhigh heat environments or when there is potential for sparking
- Available with Hansen, Schrader, Cejn, Hansen HK plug or Hansen HK socket fittings
- Can be used with AV-2000, AV-3000 or AV-3000 with SureSeal facepieces
Applications:
Maintenance work, hazardous materials handling, turn-arounds and confined space operations.Used in chemical, food processing, petrochemical, oil & gas, ship building and public utilities.
Specifications
Supplied Air Respirator System | |
Maintenance work, hazardous materials handling, turn-arounds and confi ned space operations, chemical, food processing, petrochemical, oil & gas, ship building and public utilities | |
No Facepiece | |
2216 psig |