Learn More
Scott Safety™ Supplied Air Hoses with Fittings
Description
- 3/8 in. I.D. (0.9cm) air supply hoses are constructed from rayon-braid-reinforced neoprene
Order Info
Available in various lengths and fittings
Specifications
Specifications
| Certifications/Compliance | NIOSH, MSHA |
| Model | 30010050 |
| Length (English) | 50 ft. |
| Length (Metric) | 15.2 m |