Learn More
Scott Safety™ Wescodyne™ Plus Cleaner Disinfectant
Catalog No. 19818419
Change view
Click to view available options
Volume (English):
16 oz.
1 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
19-818-419 16 oz.
19-818-420 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-818-419 Supplier Scott Safety™ Supplier No. 31001876
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
This item is not returnable. View return policy
Includes
Spray attachment
Iodine-based solution disinfects after 10 minutes of contact time
Broad spectrum Iodophor for use on facepieces, adapters, regulators.
  • Premeasured concentrate for easy mixing—just add potable water and shake
  • Spray-on application for simple disinfection
  • Kills 100% of all test microorganisms
  • Three-year shelf life in concentrate form
Certifications
EPA registered
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Kills 100% of all test microorganisms which provides one-step cleaning and disinfecting on AV-3000, AV-2000 and weld-o-vista facepieces, adapters and regulators
Includes Spray attachment
Test Time 10 min.
Volume (English) 16 oz.