Learn More
MilliporeSigma™ Supelco™ SCOTTY™ Model 206 Regulator with 1/8-inch tube connector included
Catalog No. 111003106 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Packaging Quantity:
1/Pk.
Catalog No. Packaging Quantity
11-100-3106 1/Pk.
1 options
Catalog No. 11-100-3106 Supplier MilliporeSigma™ Supelco™ Supplier No. 509965
Add to Cart
Add to Cart
Stainless steel body with 316L stainless steel/Elgiloy diaphragm. Supply pressure gauge 0-3000 psig. Delivery pressure range 0-100 psig (Delivery pressure gauge 0-200 psig). CGA 180 connector
TRUSTED_SUSTAINABILITY
Specifications
Packaging Quantity 1/Pk.
Description Syringe adapter (P/N 609010) is optional, should be purchased separately
SCOTTY is a registered trademark of Air Liquide