Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Airgas™ Transportable Pure Gases and Mixtures (48L Size) from Restek
SDP

Ideal for peak identification work

Supplier:  Restek Corporation 34472

 View more versions of this product

Catalog No. 06-802-437


Description

Description

  • CGA 165 connection and can deliver large volumes of sample
  • Scotty™ (Air Liquide) 48
  • Contents: 48L
  • Pressure: 300 psig (21 bar)
  • Outlet Fitting: CGA 165
  • Weight: 1.75 lb. (0.8kg)
  • Dimensions: 4 in. diameter × 16.25 in. height (10.2 × 41cm)
  • DOT Specifications: 39 NRC

All calibration gas standards are nonreturnable due to DOT hazardous shipping requirements

Specifications

Specifications

DOT 39 NRC
10.2 cm
Hydrogen 1% in nitrogen
Ideal for peak identification work
41 cm
1.75 lb.
0.8 kg
Hydrogen in Nitrogen
4 in.
48 L
16.25 in.
300 psi
12.68 gal.
Air Standard
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 2 Years

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.