Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558867
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558867 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558867 Supplier Invitrogen™ Supplier No. PA558867
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ILNMFRQQQK ILQQSRIVQS QRLKTIKQLY EQFIKSMEEL EKNHDNLLTG AQNEFKKEMA MLQKKIMMET QQQEIASVRK SLQSMLF Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 78%.

This gene encodes an essential structural component of the synaptonemal complex. This complex is involved in synapsis, recombination and segregation of meiotic chromosomes. Mutations in this gene are associated with azoospermia in males and susceptibility to pregnancy loss in females. Alternate splicing results in multiple transcript variants that encode the same protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCP3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Sycp3
Gene Accession No. Q8IZU3
Gene Alias Cor1; LOC615896 protein; RNASCP3; RPRGL4; Scp3; SCP-3; SPGF4; Sycp3; Synaptonemal complex protein 3; synaptonemal complex protein 3-like
Gene Symbols Sycp3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SCP3. Recombinant protein control fragment (Product #RP-97796).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50511
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.