Learn More
AnaSpec Scrambled-beta-Amyloid (1-42), 1mg
Supplier:  AnaSpec AS25383
Encompass
Aß (1-40) together with Aß (1-42) are two major C-terminal variants of the Aß protein constituting the majority of Aßs. These undergo post-secretory aggregation and deposition in the Alzheimer’s disease brain. This peptide is the scrambled sequence of Abeta 1-42.Source/Species: human
Catalog No. NC2819120
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.