Learn More
Aat Bioquest Screen Quest Calbryte-520 Probenecid-Free and Wash-Free Calcium Assay Kit (100 Plates)
SDP
Supplier:  Aat Bioquest 36319
Encompass_Preferred
Calcium flux assays are the preferred methods in drug discovery for screening G protein coupled receptors GPCR. This catalog is for 100 Plates.
Catalog No. 50-225-0178
Only null left
Add to Cart
Request a Quote
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.