Learn More
Description
- For use with Leucosep, Polystyrene and Polypropylene tubes
- Color: Red
Service Plan
No
Warranty and Services
No
Specifications
Specifications
| Product Type | Screw Cap |
| Color | White |
| For Use With (Equipment) | Leucosep, Polystyrene and Polypropylene Tubes |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.