Learn More
Thermo Scientific™ Screw Cap Micro Tubes
GREENER_CHOICE
Catalog No. 14755204 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (Metric):
0.5 mL
1.5 mL
2 mL
Bottom Shape:
Conical
Conical (Self-standing)
Non-skirted
Skirted
Closure Type:
Attached Cap
Not Included
Screw Cap with O-ring (Unattached)
Packaging:
50 tubes and attached caps/bag, 10 bags/unit
500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
500/Pack
40 product options available for selection
Product selection table with 40 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Bottom Shape Closure Type Packaging
14-755-204 0.5 mL Skirted Not Included 500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
14-755-274 0.5 mL Skirted Screw Cap with O-ring (Unattached) 500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
14-755-217 0.5 mL Skirted Attached Cap 500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
14-755-205 0.5 mL Skirted Not Included 500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
14-755-291 0.5 mL Non-skirted Not Included 500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
14-755-289 0.5 mL Non-skirted Screw Cap with O-ring (Unattached) 500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
14-755-305 0.5 mL Non-skirted Attached Cap 500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
14-755-211 0.5 mL Non-skirted Not Included 500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
21-403-540 0.5 mL Skirted Not Included 500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
21-403-203 0.5 mL Skirted Screw Cap with O-ring (Unattached) 500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
21-403-197 0.5 mL Skirted Attached Cap 500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
14-755-206 0.5 mL Skirted Not Included 500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
14-755-222 1.5 mL Non-skirted Not Included 500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
14-755-215 1.5 mL Non-skirted Screw Cap with O-ring (Unattached) 500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
14-755-208 1.5 mL Conical Attached Cap 500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
14-755-223 1.5 mL Non-skirted Not Included 500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
21-403-541 1.5 mL Skirted Not Included 500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
21-403-204 1.5 mL Skirted Screw Cap with O-ring (Unattached) 500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
14-755-287 1.5 mL Skirted Attached Cap 500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
14-755-277 1.5 mL Skirted Not Included 500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
21-403-198 1.5 mL Non-skirted Not Included 500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
21-403-196 1.5 mL Non-skirted Screw Cap with O-ring (Unattached) 500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
14-755-224 1.5 mL Non-skirted Not Included 500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
14-755-284 2 mL Skirted Not Included 500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
14-755-279 2 mL Skirted Screw Cap with O-ring (Unattached) 500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
14-755-228 2 mL Skirted Attached Cap 500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
14-755-285 2 mL Skirted Not Included 500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
14-755-233 2 mL Non-skirted Not Included 500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
14-755-226 2 mL Non-skirted Screw Cap with O-ring (Unattached) 500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
14-755-213 2 mL Non-skirted Attached Cap 500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
14-755-234 2 mL Non-skirted Not Included 500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
21-403-211 2 mL Skirted Not Included 500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
21-403-209 2 mL Skirted Screw Cap with O-ring (Unattached) 500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
21-403-201 2 mL Skirted Attached Cap 500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
14-755-286 2 mL Skirted Not Included 500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
21-403-202 2 mL Non-skirted Not Included 500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
21-403-200 2 mL Non-skirted Screw Cap with O-ring (Unattached) 500/Pack
21-403-195 2 mL Non-skirted Attached Cap 500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
14-755-235 2 mL Non-skirted Not Included 500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
21-403-199 1.5 mL Conical (Self-standing) Screw Cap with O-ring (Unattached) 50 tubes and attached caps/bag, 10 bags/unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 40 options available.
40 options
Catalog No. 14-755-204 Supplier Thermo Scientific™ Supplier No. 3422NK
Add to Cart
Edge
Add to Cart
Choose these tubes, which available in conical and free-standing designs when seeking secure, short-term sample storage in low temperatures.
Choose Thermo Scientific™ Screw Cap Micro Tubes, which available in conical and free-standing designs, when seeking secure, short-term sample storage in low temperatures. Their robust design and material selection are well suited for work with biologics and offer rich features, including ultra-clear, externally threaded tubes and UV protective amber tubes. Options include: with or without a self-standing skirted bottom, with or without tethered straps, non-knurl and knurled tubes for single-hand operation, choice of multicolored caps and volume sizes of 0.5mL, 1.5mL and 2.0mL sizes. Screw Cap Micro Tube Caps offer a durable design incorporating an O-ring that helps prevent leaks under demanding conditions.
Non-Sterile is free of Rnase/Dnase, DNA, Pyrogen & Endotoxin.
Sterile E-Beam is free of Rnase/Dnase, DNA, Pyrogen, Endotoxin, ATP, PCR Inhibitor.
Packaging is tailored to meet broad requirements. Tubes are packaged with and without caps, caps are attached or unattached, placed in a single bag and double bagged feature for clean room environments is available upon request.
100% virgin polypropylene resin specifically selected for high clarity and biologic applications
O-ring design, thickness and material selection have been optimized to provide protective features for maximum sealing ability, biologic interaction and temperature capability. The black colored O-ring is made from EPDM.
Automated System Compatible. Whether automate is used for filling, capping or labeling the product is designed to withstand high torque pressures associated with automation. Product design helps to address common automated issues like: Fissure cracks, bowl feeder jam, over/under torqued caps and cross threading.
Volume range of 0.5, 1.5 and 2.0mL.
Tube Color choices are an ultra-high clarity Natural color or the Amber option which provides ultra violet (UV) protection of 1.22% transmittance between 220 and 500nm.
Standard or Tethered cap, all available in a rainbow of colors. Visit cap product page for additional information.
Knurl tubes feature the ability to lock tube into specialty racks for single had operation.
Non knurl tubes ideal for automation and labeling, tubes are plain, removed of any labeling interference.
Skirted or Self Standing tubes can stand upright on their own, no rack required.
Conical tubes requires a rack to support them in the upright position.
Low nucleic acid and protein binding and low enzyme inhibition are features that Resin selection features.
RCF range from 18,000 to 25,000 x g.
Temperature Range -80°C to +95°C. Short term storage in -80°C, -20°C, dry ice and wet ice. Short term storage in vapor phase LN2. Suitable for a quick dip flash freeze in LN2.
Water Bath compatible up to +95°C.
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Not Autoclavable
Capacity (Metric) 0.5 mL
Color Natural
Material Polypropylene (Virgin)
Includes NonKnurl
Packaging 500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
Temperature Range (Metric) Cold Storage: -80° to +95°C. Liquid Nitrogen: Short term Vapor Phase & Quick Flash Freeze acceptable, not recommmended for LN2 submersion storage
Clean Claims Non-Sterile Free of: Rnase/Dnase, Human and Mouse DNA, Pyrogen & Endotoxin, ATP, PCR Inhibitor
Binding Property Low Nucleic Acid and Protein Binding & Low Enzyme Inhibition
Max. Torque Manual/Hand Torque: Standard: 0.125 lbsf.in to 2.5 lbsf.in; Metric: 1.412 Ncm to 28.24 Ncm. Automated/Robotic Torque Standard: 1.312 lbsf.in to 2.5 lbsf.in; Metric: 14.83 Ncm to 28.24 Ncm
Sterility Non-sterile
Graduated Yes
Closure Type Not Included
Fit for Purpose PCR Ready
Bottom Shape Skirted
Max. RCF 25,000 x g
Product Type Screw-cap Microtube
Show More Show Less
† Subject to Thermo Fisher Scientific standard limited warranty. See thermoscientific.com or your sales representative for details.
Screw cap micro tubes and caps are not recommended for autoclave. Sterile options available.
Not intended to submerge and store in liquid phase LN2.