Learn More
Bio Plas Screw Cap with O-Ring
Catalog No. 22025058
Change view
Click to view available options
Color:
Amber
Black
Blue
Clear
Green
Orange
Red
Violet
Yellow
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-025-058 Clear
22-025-062 Green
22-025-066 Amber
22-025-061 Red
22-025-059 Blue
22-025-060 Yellow
22-025-063 Orange
22-025-064 Black
22-025-065 Violet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 22-025-058 Supplier Bio Plas Supplier No. 4215R
Add to Cart
Add to Cart
Screw cap with O-ring for Bio Plas Screw Cap Microcentrifuge Tubes.
  • Screw cap with O-ring for Bio Plas Screw Cap Microcentrifuge Tubes
  • Comes in a variety of colors
Specifications
Liner Material EPDM
Product Type Screw Cap
Autoclavable Not Autoclavable
Material Polypropylene
Color Clear
Sterility Non-sterile
Disposable Single-use
Fit for Purpose Certified Quality