Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One Screw Caps for 12 mm Microcentrifuge Tubes

Catalog No. 07000530 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Natural
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07-000-530 Natural
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-000-530 Supplier Greiner Bio-One Supplier No. 366380
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with microcentrifuge tubes with 12mm diameter. Greiner Bio-One Screw Caps for Microcentrifuge Tubes is made of made of high-grade polyethylene and includes O-ring.

  • Non-sterile
  • Material: High-grade polyethylene
  • Suitable for tube-diameter: 12mm
  • With O-ring
TRUSTED_SUSTAINABILITY

Specifications

Includes O-ring
Product Type Screw Cap
Autoclavable Autoclavable
Material Polyethylene
Color Natural
No. per Case 500
Sterility Non-sterile
For Use With (Equipment) 12 mm Microcentrifuge Tubes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.